accessibility menu, dialog, popup

UserName

CAS RN 6404-29-1

IUPAC Name: 6-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Synonyms: 6-[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol): 231.29
Molecular Formula: C11H21NO4
InChi Key: RUFDYIJGNPVTAY-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCCCC(O)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)