missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-6-aminohexanoic Acid 95.0+%, TCI America™

Catalog No. p-7123443
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42755G 5 g
B427525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42755G Supplier TCI America Supplier No. B42755G
Only null left

Chemical Identifiers

CAS 6404-29-1
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.292
MDL Number MFCD00037798
InChI Key RUFDYIJGNPVTAY-UHFFFAOYSA-N
Synonym boc-epsilon-acp-oh, boc-6-aminocaproic acid, 6-tert-butoxycarbonyl amino hexanoic acid, boc-6-ahx-oh, 6-boc-amino hexanoic acid, boc-, a-acp-oh, hexanoic acid, 6-1,1-dimethylethoxy carbonyl amino, n-boc-6-aminohexanoic acid, 6-boc-amino caproic acid
PubChem CID 637602
IUPAC Name 6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCCCC(=O)O

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 166°C
Quantity 5 g
Formula Weight 231.29
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material N-(tert-Butoxycarbonyl)-6-aminohexanoic Acid
TSCA No
Recommended Storage Refrigerator