missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-6-aminohexanoic Acid 95.0+%, TCI America™
Supplier: TCI America B427525G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C11H21NO4 | |
| MFCD00037798 | |
| boc-epsilon-acp-oh, boc-6-aminocaproic acid, 6-tert-butoxycarbonyl amino hexanoic acid, boc-6-ahx-oh, 6-boc-amino hexanoic acid, boc-, a-acp-oh, hexanoic acid, 6-1,1-dimethylethoxy carbonyl amino, n-boc-6-aminohexanoic acid, 6-boc-amino caproic acid | |
| 6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
Specifications
| N-(tert-Butoxycarbonyl)-6-aminohexanoic Acid | |
| 41°C | |
| 166°C | |
| 25 g | |
| boc-epsilon-acp-oh, boc-6-aminocaproic acid, 6-tert-butoxycarbonyl amino hexanoic acid, boc-6-ahx-oh, 6-boc-amino hexanoic acid, boc-, a-acp-oh, hexanoic acid, 6-1,1-dimethylethoxy carbonyl amino, n-boc-6-aminohexanoic acid, 6-boc-amino caproic acid | |
| CC(C)(C)OC(=O)NCCCCCC(=O)O | |
| 231.292 | |
| 231.29 | |
| Crystalline Lumps |
| 6404-29-1 | |
| White-Yellow | |
| C11H21NO4 | |
| MFCD00037798 | |
| RUFDYIJGNPVTAY-UHFFFAOYSA-N | |
| 6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid | |
| 637602 | |
| ≥95.0% (GC,T) |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator