missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl N-[6-(Biotinamido)hexanoyl]-6-aminohexanoate 90.0+%, TCI America™

Supplier:  TCI America S0956100MG

 View more versions of this product

Catalog No. S0956100MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Succinimidyl N-[6-(Biotinamido)hexanoyl]-6-aminohexanoate
White
MFCD00467154
N-[6-(Biotinamido)hexanoyl]-6-aminohexanoic Acid N-Succinimidyl Ester, Biotin-LC-LC-NHS
O=C(CCCCCNC(=O)CCCCC1SCC2NC(=O)NC12)NCCCCCC(=O)ON1C(=O)CCC1=O
567.70
567.70
Crystalline Powder
89889-52-1
C26H41N5O7S
100 mg
ATYCFNRXENKXSE-UHFFFAOYNA-N
2,5-dioxopyrrolidin-1-yl 6-[6-(5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamido)hexanamido]hexanoate
16219024
≥95.0% (HPLC)

Chemical Identifiers

89889-52-1
567.70
ATYCFNRXENKXSE-UHFFFAOYNA-N
16219024
O=C(CCCCCNC(=O)CCCCC1SCC2NC(=O)NC12)NCCCCCC(=O)ON1C(=O)CCC1=O
C26H41N5O7S
MFCD00467154
N-[6-(Biotinamido)hexanoyl]-6-aminohexanoic Acid N-Succinimidyl Ester, Biotin-LC-LC-NHS
2,5-dioxopyrrolidin-1-yl 6-[6-(5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamido)hexanamido]hexanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions