accessibility menu, dialog, popup

UserName

CAS RN 89889-52-1

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 6-[6-(5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamido)hexanamido]hexanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 6-[6-(5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamido)hexanamido]hexanoate
Molecular Weight (g/mol): 567.7
Molecular Formula: C26H41N5O7S
InChi Key: ATYCFNRXENKXSE-UHFFFAOYNA-N
SMILES: O=C(CCCCCNC(=O)CCCCC1SCC2NC(=O)NC12)NCCCCCC(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results