missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl N-[6-(Biotinamido)hexanoyl]-6-aminohexanoate 90.0+%, TCI America™
$285.21 - $859.33
Chemical Identifiers
| CAS | 89889-52-1 |
|---|---|
| Molecular Formula | C26H41N5O7S |
| Molecular Weight (g/mol) | 567.70 |
| MDL Number | MFCD00467154 |
| InChI Key | ATYCFNRXENKXSE-UHFFFAOYNA-N |
| Synonym | N-[6-(Biotinamido)hexanoyl]-6-aminohexanoic Acid N-Succinimidyl Ester, Biotin-LC-LC-NHS |
| PubChem CID | 16219024 |
| IUPAC Name | 2,5-dioxopyrrolidin-1-yl 6-[6-(5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamido)hexanamido]hexanoate |
| SMILES | O=C(CCCCCNC(=O)CCCCC1SCC2NC(=O)NC12)NCCCCCC(=O)ON1C(=O)CCC1=O |
Chemical Identifiers
| 89889-52-1 | |
| 567.70 | |
| ATYCFNRXENKXSE-UHFFFAOYNA-N | |
| 16219024 | |
| O=C(CCCCCNC(=O)CCCCC1SCC2NC(=O)NC12)NCCCCCC(=O)ON1C(=O)CCC1=O |
| C26H41N5O7S | |
| MFCD00467154 | |
| N-[6-(Biotinamido)hexanoyl]-6-aminohexanoic Acid N-Succinimidyl Ester, Biotin-LC-LC-NHS | |
| 2,5-dioxopyrrolidin-1-yl 6-[6-(5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamido)hexanamido]hexanoate |
Specifications
| 89889-52-1 | |
| C26H41N5O7S | |
| 25 mg | |
| ATYCFNRXENKXSE-UHFFFAOYNA-N | |
| 2,5-dioxopyrrolidin-1-yl 6-[6-(5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamido)hexanamido]hexanoate | |
| 16219024 | |
| ≥95.0% (HPLC) | |
| N-Succinimidyl N-[6-(Biotinamido)hexanoyl]-6-aminohexanoate |
| White | |
| MFCD00467154 | |
| N-[6-(Biotinamido)hexanoyl]-6-aminohexanoic Acid N-Succinimidyl Ester, Biotin-LC-LC-NHS | |
| O=C(CCCCCNC(=O)CCCCC1SCC2NC(=O)NC12)NCCCCCC(=O)ON1C(=O)CCC1=O | |
| 567.70 | |
| 567.70 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer