missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 4-Benzoylbenzoate 98.0+%, TCI America™

Catalog No. p-7135970
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0863200MG 200 mg
S08631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0863200MG Supplier TCI America Supplier No. S0863200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91990-88-4
Molecular Formula C18H13NO5
Molecular Weight (g/mol) 323.30
MDL Number MFCD00058572
InChI Key MVQNJLJLEGZFGP-UHFFFAOYSA-N
Synonym 4-Benzoylbenzoic Acid N-Succinimidyl Ester
PubChem CID 2762595
IUPAC Name 2,5-dioxopyrrolidin-1-yl 4-benzoylbenzoate
SMILES O=C(ON1C(=O)CCC1=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 209°C
Color White
Quantity 200 mg
Formula Weight 323.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 4-Benzoylbenzoate
TSCA No
Recommended Storage Refrigerator