missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals J64834MC

 View more versions of this product

Catalog No. AAJ64834MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate is an amine reactive reagent for azide incorporation. It contains a PEG spacer.

Solubility
Soluble in dimethyl sulfoxide and acetonitrile.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate
C15H24N4O8
100mg
Soluble in dimethyl sulfoxide and acetonitrile.
944251-24-5
MFCD13184948
Azido-PEG4-NHS ester
388.37

Chemical Identifiers

944251-24-5
C15H24N4O8
MFCD13184948
Azido-PEG4-NHS ester
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only