CAS RN 944251-24-5
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 1-azido-3,6,9,12-tetraoxapentadecan-15-oate
Synonyms:
2,5-dioxopyrrolidin-1-yl 1-azido-3,6,9,12-tetraoxapentadecan-15-oate
Molecular Weight (g/mol):
388.38
Molecular Formula:
C15H24N4O8
InChi Key:
OIGKWPIMJCPGGD-UHFFFAOYSA-N
SMILES:
[N-]=[N+]=NCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O
1
–
2
of
2
results
1
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%, Thermo Scientific™
CAS: 944251-24-5 Molecular Formula: C15H24N4O8 MDL Number: MFCD13184948 Synonym: Azido-PEG4-NHS ester
2
Azido-PEG4-NHS Ester, TCI America™
CAS: 944251-24-5 Molecular Formula: C15H24N4O8 Molecular Weight (g/mol): 388.377 InChI Key: OIGKWPIMJCPGGD-UHFFFAOYSA-N Synonym: NHS-PEG4-Azide, N-Succinimidyl 15-Azido-4,7,10,13-tetraoxapentadecanoate, 15-Azido-4,7,10,13-tetraoxapentadecanoic Acid N-Succinimidyl Ester PubChem CID: 51340931 IUPAC Name: (2,5-dioxopyrrolidin-1-yl) 3-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethoxy]propanoate SMILES: C1CC(=O)N(C1=O)OC(=O)CCOCCOCCOCCOCCN=[N+]=[N-]