missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%, Thermo Scientific™

Chemical Identifiers

CAS 944251-24-5
Molecular Formula C15H24N4O8
Molecular Weight (g/mol) 388.38
MDL Number MFCD13184948
InChI Key OIGKWPIMJCPGGD-UHFFFAOYSA-N
Synonym Azido-PEG4-NHS ester
IUPAC Name 2,5-dioxopyrrolidin-1-yl 1-azido-3,6,9,12-tetraoxapentadecan-15-oate
SMILES [N-]=[N+]=NCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ64834MB
View Documents
N/A 25mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ64834MC
View Documents
N/A 100mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate is an amine reactive reagent for azide incorporation. It contains a PEG spacer.

Solubility
Soluble in dimethyl sulfoxide and acetonitrile.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

944251-24-5
388.38
OIGKWPIMJCPGGD-UHFFFAOYSA-N
2,5-dioxopyrrolidin-1-yl 1-azido-3,6,9,12-tetraoxapentadecan-15-oate
C15H24N4O8
MFCD13184948
Azido-PEG4-NHS ester
[N-]=[N+]=NCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O

Specifications

944251-24-5
MFCD13184948
Soluble in dimethyl sulfoxide and acetonitrile.
[N-]=[N+]=NCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O
388.38
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate
C15H24N4O8
Azido-PEG4-NHS ester
OIGKWPIMJCPGGD-UHFFFAOYSA-N
2,5-dioxopyrrolidin-1-yl 1-azido-3,6,9,12-tetraoxapentadecan-15-oate
388.37
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only