missing translation for 'onlineSavingsMsg'
Learn More

N,S-Dimethylisothiouronium hydriodide, 98%

Catalog No. AAH2743914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2743914 25 g
AAH2743906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2743914 Supplier Thermo Scientific Chemicals Supplier No. H2743914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41306-45-0 | C3H9IN2S | 232.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41306-45-0
Molecular Formula C3H9IN2S
Molecular Weight (g/mol) 232.083
MDL Number MFCD00192092
InChI Key UDUMWDDMSHXGGQ-UHFFFAOYSA-N
Synonym 1,2-dimethyl-2-thiopseudourea hydriodide, 1,2-dimethylisothiourea hydriodate, n,s-dimethyl-isothiourea hydroiodide, 1,2-dimethyl-isothiourea hydroiodide, n,s-dimethylisothiouronium hydriodide, iminomethylthiomethyl methylamine, iodide, 1,2-dimethyl-2-thiopseudoureahydriodide, n-methyl methylsulfanyl methanimidamide hydroiodide, n'-methyl methylsulfanyl methanimidamide hydroiodide
PubChem CID 12263909
IUPAC Name methyl N'-methylcarbamimidothioate;hydroiodide
SMILES CN=C(N)SC.I

Specifications

Melting Point 134°C to 136°C
Quantity 25 g
Sensitivity Hygroscopic; Light sensitive
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material N,S-Dimethylisothiouronium hydriodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only