accessibility menu, dialog, popup

UserName

CAS RN 41306-45-0

IUPAC Name: N'-methyl(methylsulfanyl)methanimidamide hydroiodide
Synonyms: N'-methylmethylsulfanylmethanimidamide hydroiodide
Molecular Weight (g/mol): 232.08
Molecular Formula: C3H9IN2S
InChi Key: UDUMWDDMSHXGGQ-UHFFFAOYSA-N
SMILES: I.CSC(N)=NC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results