missing translation for 'onlineSavingsMsg'
Learn More

n-Propylmagnesium chloride, 1M in MeTHF

CAS: 2234-82-4 | C3H7ClMg | 102.84 g/mol

$124.62 - $466.04

Chemical Identifiers

CAS 2234-82-4
Molecular Formula C3H7ClMg
Molecular Weight (g/mol) 102.84
MDL Number MFCD00000474
InChI Key AMXVKMXLDJXAPM-UHFFFAOYSA-M
IUPAC Name chloro(propyl)magnesium
SMILES CCC[Mg]Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH51162AE
View Documents
Thermo Scientific Chemicals
H51162AE
100 mL
Each for $124.62
Only null left
 
AAH51162AP
View Documents
Thermo Scientific Chemicals
H51162AP
500 mL
Each for $466.04
Only null left
 
Description

Description

Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). Also used as reactant in Grignard-reactions for the introduction of the n-propyl group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). Also used as reactant in Grignard-reactions for the introduction of the n-propyl group.

Solubility
Reacts with water.

Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2234-82-4
102.84
AMXVKMXLDJXAPM-UHFFFAOYSA-M
CCC[Mg]Cl
C3H7ClMg
MFCD00000474
chloro(propyl)magnesium

Specifications

2234-82-4
MFCD00000474
UN3399
Reacts with water.
CCC[Mg]Cl
102.84
1M in MeTHF
C3H7ClMg
100 mL
Air and moisture sensitive
AMXVKMXLDJXAPM-UHFFFAOYSA-M
chloro(propyl)magnesium
102.85
n-Propylmagnesium chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H225-H261-H302-H314-H335

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only