accessibility menu, dialog, popup

UserName

CAS RN 2234-82-4

CH3MgCl

CAS RN 2234-82-4

IUPAC Name: chloro(propyl)magnesium
Synonyms: propylmagnesium chloride
Molecular Weight (g/mol): 102.84
Molecular Formula: C3H7ClMg
InChi Key: AMXVKMXLDJXAPM-UHFFFAOYSA-M
SMILES: CCC[Mg]Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results