Learn More
n-Propylmagnesium chloride, 1M in MeTHF
CAS: 2234-82-4 | C3H7ClMg | 102.84 g/mol
Supplier: Thermo Scientific Chemicals H51162AP
Description
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). Also used as reactant in Grignard-reactions for the introduction of the n-propyl group.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGrignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). Also used as reactant in Grignard-reactions for the introduction of the n-propyl group.
Solubility
Reacts with water.
Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| n-Propylmagnesium chloride | |
| C3H7ClMg | |
| 500 mL | |
| Air and moisture sensitive | |
| AMXVKMXLDJXAPM-UHFFFAOYSA-M | |
| chloro(propyl)magnesium | |
| 102.85 |
| 2234-82-4 | |
| MFCD00000474 | |
| UN3399 | |
| Reacts with water. | |
| CCC[Mg]Cl | |
| 102.84 | |
| 1M in MeTHF |
Chemical Identifiers
| 2234-82-4 | |
| 102.84 | |
| AMXVKMXLDJXAPM-UHFFFAOYSA-M | |
| CCC[Mg]Cl |
| C3H7ClMg | |
| MFCD00000474 | |
| chloro(propyl)magnesium |
Safety and Handling
GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H261-H302-H314-H335
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only