missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylurethane, 98%

Catalog No. p-7044176
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0502714 25 g
AAL0502722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0502714 Supplier Thermo Scientific Chemicals Supplier No. L0502714
Only null left

CAS: 101-99-5 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101-99-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00026806
InChI Key LBKPGNUOUPTQKA-UHFFFAOYSA-N
Synonym n-phenylurethane, phenylurethane, ethyl phenylcarbamate, phenylurethan, ethyl carbanilate, euphorin, keimstop, urethane, phenyl, carbanilic acid, ethyl ester, phenylethyl carbamate
PubChem CID 7591
IUPAC Name ethyl N-phenylcarbamate
SMILES CCOC(=O)NC1=CC=CC=C1

Specifications

Melting Point 49°C to 51°C
Density 1.106
Boiling Point 237°C
Refractive Index 1.5375
Quantity 25 g
Beilstein 1942785
Merck Index 14,7320
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material N-Phenylurethane
Hazard Category H351
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 202-995-9
RTECSNumber FD8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only