missing translation for 'onlineSavingsMsg'
Learn More

N-(Phenylthio)phthalimide 98.0+%, TCI America™

Catalog No. P12545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12545G 5 g
P125425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12545G Supplier TCI America Supplier No. P12545G
Only null left

Chemical Identifiers

CAS 14204-27-4
Molecular Formula C14H9NO2S
Molecular Weight (g/mol) 255.29
MDL Number MFCD00192396
InChI Key NMHKBABHRKQHOL-UHFFFAOYSA-N
PubChem CID 5127161
IUPAC Name 2-(phenylsulfanyl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1N(SC2=CC=CC=C2)C(=O)C2=CC=CC=C12

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 5 g
Formula Weight 255.29
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(Phenylthio)phthalimide
EINECSNumber (5)-0090
TSCA No