accessibility menu, dialog, popup

UserName

CAS RN 14204-27-4

SNOO

CAS RN 14204-27-4

IUPAC Name: 2-(phenylsulfanyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(phenylsulfanyl)isoindole-1,3-dione
Molecular Weight (g/mol): 255.29
Molecular Formula: C14H9NO2S
InChi Key: NMHKBABHRKQHOL-UHFFFAOYSA-N
SMILES: O=C1N(SC2=CC=CC=C2)C(=O)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results