missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyliminodiacetic acid, 97%

CAS: 1137-73-1 | C10H11NO4 | 209.201 g/mol

$145.81 - $541.95

Chemical Identifiers

CAS 1137-73-1
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
MDL Number MFCD00043570
InChI Key GQBWTAGIANQVGB-UHFFFAOYSA-N
Synonym n-phenyliminodiacetic acid, glycine, n-carboxymethyl-n-phenyl, acetic acid,1-phenyliminodi, 2,2'-phenylazanediyl diacetic acid, 2,2'-phenylimino diacetic acid, n-carboxymethyl-n-phenylglycine, n,n-anilinediacetic acid, n-phenyliminodiacetic acid anhydride, nn-di carboxymethyl aniline, glycine,n-carboxymethyl-n-phenyl
PubChem CID 136915
IUPAC Name 2-[N-(carboxymethyl)anilino]acetic acid
SMILES C1=CC=C(C=C1)N(CC(=O)O)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0843706
View Documents
Thermo Scientific Chemicals
L0843706
5 g
Each for $145.81
Only null left
 
AAL0843714
View Documents
Thermo Scientific Chemicals
L0843714
25 g
Each for $541.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1137-73-1
209.201
GQBWTAGIANQVGB-UHFFFAOYSA-N
136915
C1=CC=C(C=C1)N(CC(=O)O)CC(=O)O
C10H11NO4
MFCD00043570
n-phenyliminodiacetic acid, glycine, n-carboxymethyl-n-phenyl, acetic acid,1-phenyliminodi, 2,2'-phenylazanediyl diacetic acid, 2,2'-phenylimino diacetic acid, n-carboxymethyl-n-phenylglycine, n,n-anilinediacetic acid, n-phenyliminodiacetic acid anhydride, nn-di carboxymethyl aniline, glycine,n-carboxymethyl-n-phenyl
2-[N-(carboxymethyl)anilino]acetic acid

Specifications

1137-73-1
C10H11NO4
5 g
n-phenyliminodiacetic acid, glycine, n-carboxymethyl-n-phenyl, acetic acid,1-phenyliminodi, 2,2'-phenylazanediyl diacetic acid, 2,2'-phenylimino diacetic acid, n-carboxymethyl-n-phenylglycine, n,n-anilinediacetic acid, n-phenyliminodiacetic acid anhydride, nn-di carboxymethyl aniline, glycine,n-carboxymethyl-n-phenyl
C1=CC=C(C=C1)N(CC(=O)O)CC(=O)O
209.201
209.2
N-Phenyliminodiacetic acid
∽140°C (decomposition)
MFCD00043570
2808435
GQBWTAGIANQVGB-UHFFFAOYSA-N
2-[N-(carboxymethyl)anilino]acetic acid
136915
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only