missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyliminodiacetic acid, 97%

CAS: 1137-73-1 | C10H11NO4 | 209.201 g/mol

Supplier:  Thermo Scientific Chemicals L0843714

 View more versions of this product

Catalog No. AAL0843714


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Phenyliminodiacetic acid
∽140°C (decomposition)
MFCD00043570
2808435
GQBWTAGIANQVGB-UHFFFAOYSA-N
2-[N-(carboxymethyl)anilino]acetic acid
136915
97%
1137-73-1
C10H11NO4
25 g
n-phenyliminodiacetic acid, glycine, n-carboxymethyl-n-phenyl, acetic acid,1-phenyliminodi, 2,2'-phenylazanediyl diacetic acid, 2,2'-phenylimino diacetic acid, n-carboxymethyl-n-phenylglycine, n,n-anilinediacetic acid, n-phenyliminodiacetic acid anhydride, nn-di carboxymethyl aniline, glycine,n-carboxymethyl-n-phenyl
C1=CC=C(C=C1)N(CC(=O)O)CC(=O)O
209.201
209.2

Chemical Identifiers

1137-73-1
209.201
GQBWTAGIANQVGB-UHFFFAOYSA-N
136915
C1=CC=C(C=C1)N(CC(=O)O)CC(=O)O
C10H11NO4
MFCD00043570
n-phenyliminodiacetic acid, glycine, n-carboxymethyl-n-phenyl, acetic acid,1-phenyliminodi, 2,2'-phenylazanediyl diacetic acid, 2,2'-phenylimino diacetic acid, n-carboxymethyl-n-phenylglycine, n,n-anilinediacetic acid, n-phenyliminodiacetic acid anhydride, nn-di carboxymethyl aniline, glycine,n-carboxymethyl-n-phenyl
2-[N-(carboxymethyl)anilino]acetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only