missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylacrylamide 98.0+%, TCI America™
Supplier: TCI America P209325G
Specifications
| N-Phenylacrylamide | |
| 105°C | |
| C9H9NO | |
| 25 g | |
| C=CC(=O)NC1=CC=CC=C1 | |
| 147.18 | |
| 147.18 | |
| Crystalline Powder |
| 2210-24-4 | |
| White | |
| MFCD00080617 | |
| BPCNEKWROYSOLT-UHFFFAOYSA-N | |
| N-phenylprop-2-enamide | |
| 221792 | |
| ≥98.0% (GC,N) |
Chemical Identifiers
| 2210-24-4 | |
| 147.18 | |
| BPCNEKWROYSOLT-UHFFFAOYSA-N | |
| N-phenylprop-2-enamide |
| C9H9NO | |
| MFCD00080617 | |
| 221792 | |
| C=CC(=O)NC1=CC=CC=C1 |
Safety and Handling
TSCA : No