missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylacrylamide 98.0+%, TCI America™

Catalog No. p-7135402
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
P20935G 5 g
P209325G 25 g
2 options

Catalog No. P20935G

Supplier: TCI America P20935G

Only null left

Chemical Identifiers

CAS 2210-24-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00080617
InChI Key BPCNEKWROYSOLT-UHFFFAOYSA-N
PubChem CID 221792
IUPAC Name N-phenylprop-2-enamide
SMILES C=CC(=O)NC1=CC=CC=C1

Specifications

Melting Point 105°C
Color White
Quantity 5 g
Formula Weight 147.18
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylacrylamide