missing translation for 'onlineSavingsMsg'
Learn More

n-Pentyl benzoate, 98+%

Catalog No. p-7080596
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1017722 100 g
AAA1017736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1017722 Supplier Thermo Scientific Chemicals Supplier No. A1017722
Only null left

CAS: 2049-96-9 | C12H16O2 | 192.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2049-96-9
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00048843
InChI Key QKNZNUNCDJZTCH-UHFFFAOYSA-N
Synonym amyl benzoate, n-pentyl benzoate, benzoic acid, pentyl ester, n-amyl benzoate, unii-cle7k4b29s, cle7k4b29s, amylbenzoat, amylbenzoate, n amyl benxoate, benzoic acid pentyl
PubChem CID 16296
IUPAC Name pentyl benzoate
SMILES CCCCCOC(=O)C1=CC=CC=C1

Specifications

Density 0.99
Boiling Point 260°C
Flash Point 130°C (266°F)
Refractive Index 1.495
Quantity 100 g
Formula Weight 192.26
Percent Purity ≥98%
Chemical Name or Material n-Pentyl benzoate
EINECSNumber 218-077-6
RTECSNumber DH6276000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only