accessibility menu, dialog, popup

UserName

CAS RN 2049-96-9

IUPAC Name: pentyl benzoate
Synonyms: amyl benzoate
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: QKNZNUNCDJZTCH-UHFFFAOYSA-N
SMILES: CCCCCOC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)