missing translation for 'onlineSavingsMsg'
Learn More

N-(p-Tolyl)-2-naphthylamine 98.0+%, TCI America™

Catalog No. t2065500mg
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2065500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2065500MG Supplier TCI America Supplier No. T2065500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 644-16-6
Molecular Formula C17H15N
Molecular Weight (g/mol) 233.314
InChI Key IBJHDUPUTZQCLL-UHFFFAOYSA-N
Synonym 2-(p-Toluidino)naphthalene
PubChem CID 229322
IUPAC Name N-(4-methylphenyl)naphthalen-2-amine
SMILES CC1=CC=C(C=C1)NC2=CC3=CC=CC=C3C=C2

Specifications

Color Yellow
Quantity 500 mg
Formula Weight 233.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(p-Tolyl)-2-naphthylamine