accessibility menu, dialog, popup

UserName

CH3HN

CAS RN 644-16-6

IUPAC Name: N-(4-methylphenyl)naphthalen-2-amine
Synonyms: N-(4-methylphenyl)naphthalen-2-amine
Molecular Weight (g/mol): 233.31
Molecular Formula: C17H15N
InChi Key: IBJHDUPUTZQCLL-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC2=CC=C3C=CC=CC3=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results