missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl Phenyl Ether 98.0+%, TCI America™

Catalog No. o022410ml
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O022410ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O022410ML Supplier TCI America Supplier No. O022410ML

Chemical Identifiers

CAS 1818-07-1
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.329
MDL Number MFCD00191564
InChI Key ZPIRTVJRHUMMOI-UHFFFAOYSA-N
Synonym n-Octyloxybenzene
PubChem CID 74558
IUPAC Name octoxybenzene
SMILES CCCCCCCCOC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 134°C
Quantity 10 mL
Formula Weight 206.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octyl Phenyl Ether