accessibility menu, dialog, popup

UserName

CAS RN 1818-07-1

IUPAC Name: (octyloxy)benzene
Synonyms: benzene, (octyloxy)-
Molecular Weight (g/mol): 206.33
Molecular Formula: C14H22O
InChi Key: ZPIRTVJRHUMMOI-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results