missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ n-Octyl-β-D-glucopyranoside, Calbiochem™,

Supplier:  MilliporeSigma™ 4944591GM

Quantity 1 g
Packaging Glass Bottle

 View more versions of this product

Catalog No. 4944591GM


Only null left

Description

Description

Specifications

Chemical Identifiers

29836-26-8
292.37
HEGSGKPQLMEBJL-RKQHYHRCSA-N
62852
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
C14H28O6
MFCD00063288
OG, n-Octylglucoside
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol

Specifications

White
1 g
C14H28O6
OG, n-Octylglucoside
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
292.37
≥98% (HPLC)
29836-26-8
Glass Bottle
MFCD00063288
HEGSGKPQLMEBJL-RKQHYHRCSA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol
62852
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions