missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ n-Octyl-β-D-glucopyranoside, Calbiochem™,

Catalog No. p-7082971
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4944595GM 5 g Glass Bottle
49445925GM 25 g Glass Bottle
494459100GM 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 4944595GM Supplier MilliporeSigma™ Supplier No. 4944595GM

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
Synonym OG, n-Octylglucoside
PubChem CID 62852
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Quantity 5 g
Packaging Glass Bottle
Percent Purity ≥98% (HPLC)