missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ n-Octyl-β-D-glucopyranoside, Calbiochem™,
Supplier: MilliporeSigma™ 494459100GM
| Quantity | 100 g |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| 29836-26-8 | |
| 292.37 | |
| HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
| 62852 | |
| CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O |
| C14H28O6 | |
| MFCD00063288 | |
| OG, n-Octylglucoside | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol |
Specifications
| White | |
| 100 g | |
| C14H28O6 | |
| OG, n-Octylglucoside | |
| CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O | |
| 292.37 | |
| ≥98% (HPLC) |
| 29836-26-8 | |
| Glass Bottle | |
| MFCD00063288 | |
| HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol | |
| 62852 |