missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl 4-hydroxybenzoate, 98%

CAS: 1219-38-1 | C15H22O3 | 250.34 g/mol

Supplier:  Thermo Scientific Chemicals A1596414

 View more versions of this product

Catalog No. AAA1596414


Only null left
Explore available promotions

Description

Description

n-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in alcohol, phenoxy ethanol and propylene glycol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

n-Octyl 4-hydroxybenzoate
50°C to 53°C
MFCD00016482
octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german
RIKCMEDSBFQFAL-UHFFFAOYSA-N
octyl 4-hydroxybenzoate
14642
98%
1219-38-1
C15H22O3
25 g
Soluble in alcohol,phenoxy ethanol and propylene glycol. Insoluble in water.
CCCCCCCCOC(=O)C1=CC=C(O)C=C1
250.34
250.34

Chemical Identifiers

1219-38-1
250.34
RIKCMEDSBFQFAL-UHFFFAOYSA-N
14642
CCCCCCCCOC(=O)C1=CC=C(O)C=C1
C15H22O3
MFCD00016482
octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german
octyl 4-hydroxybenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-943-2

RTECSNumber : DH2529050

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only