accessibility menu, dialog, popup

UserName

CAS RN 1219-38-1

IUPAC Name: octyl 4-hydroxybenzoate
Synonyms: octyl 4-hydroxybenzoate
Molecular Weight (g/mol): 250.34
Molecular Formula: C15H22O3
InChi Key: RIKCMEDSBFQFAL-UHFFFAOYSA-N
SMILES: CCCCCCCCOC(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)