missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octyl 4-Hydroxybenzoate 98.0+%, TCI America™
$154.02 - $154.02
Chemical Identifiers
| CAS | 1219-38-1 |
|---|---|
| Molecular Formula | C15H22O3 |
| Molecular Weight (g/mol) | 250.34 |
| MDL Number | MFCD00016482 |
| InChI Key | RIKCMEDSBFQFAL-UHFFFAOYSA-N |
| Synonym | octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german |
| PubChem CID | 14642 |
| IUPAC Name | octyl 4-hydroxybenzoate |
| SMILES | CCCCCCCCOC(=O)C1=CC=C(O)C=C1 |
Chemical Identifiers
| 1219-38-1 | |
| 250.34 | |
| RIKCMEDSBFQFAL-UHFFFAOYSA-N | |
| 14642 | |
| CCCCCCCCOC(=O)C1=CC=C(O)C=C1 |
| C15H22O3 | |
| MFCD00016482 | |
| octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german | |
| octyl 4-hydroxybenzoate |
Specifications
| 1219-38-1 | |
| White | |
| C15H22O3 | |
| 25 g | |
| RIKCMEDSBFQFAL-UHFFFAOYSA-N | |
| octyl 4-hydroxybenzoate | |
| 14642 | |
| ≥98.0% (GC,T) | |
| n-Octyl 4-Hydroxybenzoate |
| 53°C | |
| 172°C | |
| MFCD00016482 | |
| octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german | |
| CCCCCCCCOC(=O)C1=CC=C(O)C=C1 | |
| 250.34 | |
| 250.34 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1585
RTECSNumber : DH2529050
TSCA : No