missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl 4-Hydroxybenzoate 98.0+%, TCI America™

Catalog No. O044325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O044325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O044325G Supplier TCI America Supplier No. O044325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1219-38-1
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number MFCD00016482
InChI Key RIKCMEDSBFQFAL-UHFFFAOYSA-N
Synonym octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german
PubChem CID 14642
IUPAC Name octyl 4-hydroxybenzoate
SMILES CCCCCCCCOC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 53°C
Color White
Boiling Point 172°C
Quantity 25 g
Formula Weight 250.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material n-Octyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
RTECSNumber DH2529050
TSCA No