missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octanamide 98.0+%, TCI America™
$227.41 - $361.62
Chemical Identifiers
| CAS | 629-01-6 |
|---|---|
| Molecular Formula | C8H17NO |
| Molecular Weight (g/mol) | 143.23 |
| MDL Number | MFCD00191675 |
| InChI Key | LTHCSWBWNVGEFE-UHFFFAOYSA-N |
| PubChem CID | 69414 |
| IUPAC Name | octanamide |
| SMILES | CCCCCCCC(=O)N |