accessibility menu, dialog, popup

UserName

H3CNH2O

CAS RN 629-01-6

IUPAC Name: octanamide
Synonyms: octamide
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: LTHCSWBWNVGEFE-UHFFFAOYSA-N
SMILES: CCCCCCCC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results