missing translation for 'onlineSavingsMsg'
Learn More

n-Octanamide 98.0+%, TCI America™

Supplier:  TCI America O023825G

 View more versions of this product

Catalog No. O023825G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

n-Octanamide
105°C
C8H17NO
25 g
CCCCCCCC(=O)N
143.23
143.23
Crystalline Powder
629-01-6
White
MFCD00191675
LTHCSWBWNVGEFE-UHFFFAOYSA-N
octanamide
69414
≥98.0% (GC)

Chemical Identifiers

629-01-6
143.23
LTHCSWBWNVGEFE-UHFFFAOYSA-N
octanamide
C8H17NO
MFCD00191675
69414
CCCCCCCC(=O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0824

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions