missing translation for 'onlineSavingsMsg'
Learn More

N,N'-p-Phenylenebisacetamide, 98%

Catalog No. p-7044820
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0838106 5 g
AAL0838114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0838106 Supplier Thermo Scientific Chemicals Supplier No. L0838106
Only null left

CAS: 140-50-1 | C10H12N2O2 | 192.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140-50-1
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.218
MDL Number MFCD00026142
InChI Key KVEDKKLZCJBVNP-UHFFFAOYSA-N
Synonym 1,4-diacetamidobenzene, n,n'-diacetyl-1,4-phenylenediamine, n,n'-p-phenylenebisacetamide, p-phenylenediacetamide, n,n'-diacetyl-p-phenylenediamine, 4'-acetamidoacetanilide, 1,4-bisacetamidobenzene, acetamide, n,n'-1,4-phenylenebis, n-4-acetamidophenyl acetamide, n,n'-p-phenylene di acetamide
PubChem CID 67324
IUPAC Name N-(4-acetamidophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)NC(=O)C

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 2806271
Formula Weight 192.22
Percent Purity 98%
Chemical Name or Material N,N'-p-Phenylenebisacetamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-417-3
RTECSNumber AC7709500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only