accessibility menu, dialog, popup

UserName

CH3ONHNHCH3O

CAS RN 140-50-1

IUPAC Name: N-(4-acetamidophenyl)acetamide
Synonyms: N-(4-acetamidophenyl)acetamide
Molecular Weight (g/mol): 192.22
Molecular Formula: C10H12N2O2
InChi Key: KVEDKKLZCJBVNP-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(NC(C)=O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)