missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diphenylacetamide 98.0+%, TCI America™

Catalog No. D086825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D086825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D086825G Supplier TCI America Supplier No. D086825G
Only null left

Chemical Identifiers

CAS 519-87-9
Molecular Formula C14H13NO
Molecular Weight (g/mol) 211.26
MDL Number MFCD00008685
InChI Key DKLYDESVXZKCFI-UHFFFAOYSA-N
Synonym N-Acetyldiphenylamine
PubChem CID 10615
IUPAC Name N,N-diphenylacetamide
SMILES CC(=O)N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 25 g
Formula Weight 211.26
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diphenylacetamide
RTECSNumber AB8133000
TSCA Yes