accessibility menu, dialog, popup

UserName

CAS RN 519-87-9

H3CON

CAS RN 519-87-9

IUPAC Name: N,N-diphenylacetamide
Synonyms: N,N-diphenylacetamide
Molecular Weight (g/mol): 211.26
Molecular Formula: C14H13NO
InChi Key: DKLYDESVXZKCFI-UHFFFAOYSA-N
SMILES: CC(=O)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results