missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-m-toluidine, 97+%

CAS: 121-72-2 | C9H13N | 135.21 g/mol

$204.78 - $204.78

Chemical Identifiers

CAS 121-72-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008305
InChI Key CWOMTHDOJCARBY-UHFFFAOYSA-N
Synonym n,n-dimethyl-m-toluidine, benzenamine, n,n,3-trimethyl, dimethyl-m-toluidine, m-methyl-n,n-dimethylaniline, n,n-dimethyl-m-methylaniline, m,n,n-trimethylaniline, n,n-dimethyl-3-methylaniline, dimetil-m-toluidina, n,n,3-trimethylbenzenamine, benzene, 1-dimethylamino-3-methyl
PubChem CID 8488
IUPAC Name N,N,3-trimethylaniline
SMILES CN(C)C1=CC=CC(C)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1332622
View Documents
Thermo Scientific Chemicals
A1332622
100 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-72-2
135.21
CWOMTHDOJCARBY-UHFFFAOYSA-N
8488
CN(C)C1=CC=CC(C)=C1
C9H13N
MFCD00008305
n,n-dimethyl-m-toluidine, benzenamine, n,n,3-trimethyl, dimethyl-m-toluidine, m-methyl-n,n-dimethylaniline, n,n-dimethyl-m-methylaniline, m,n,n-trimethylaniline, n,n-dimethyl-3-methylaniline, dimetil-m-toluidina, n,n,3-trimethylbenzenamine, benzene, 1-dimethylamino-3-methyl
N,N,3-trimethylaniline

Specifications

121-72-2
0.932
85°C (185°F)
1.55
100 g
1422766
CWOMTHDOJCARBY-UHFFFAOYSA-N
N,N,3-trimethylaniline
8488
98%
-15°C
212°C to 215°C
C9H13N
MFCD00008305
UN1708
n,n-dimethyl-m-toluidine, benzenamine, n,n,3-trimethyl, dimethyl-m-toluidine, m-methyl-n,n-dimethylaniline, n,n-dimethyl-m-methylaniline, m,n,n-trimethylaniline, n,n-dimethyl-3-methylaniline, dimetil-m-toluidina, n,n,3-trimethylbenzenamine, benzene, 1-dimethylamino-3-methyl
CN(C)C1=CC=CC(C)=C1
135.21
135.21
N,N-Dimethyl-m-toluidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H373-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c

H301+H311+H331-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUIDINES, LIQUID

EINECSNumber : 204-495-6

RTECSNumber : XU5798000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only