missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-m-toluidine, 97+%

Catalog No. aaa1332622
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1332622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1332622 Supplier Thermo Scientific Chemicals Supplier No. A1332622
Only null left

CAS: 121-72-2 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-72-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008305
InChI Key CWOMTHDOJCARBY-UHFFFAOYSA-N
Synonym n,n-dimethyl-m-toluidine, benzenamine, n,n,3-trimethyl, dimethyl-m-toluidine, m-methyl-n,n-dimethylaniline, n,n-dimethyl-m-methylaniline, m,n,n-trimethylaniline, n,n-dimethyl-3-methylaniline, dimetil-m-toluidina, n,n,3-trimethylbenzenamine, benzene, 1-dimethylamino-3-methyl
PubChem CID 8488
IUPAC Name N,N,3-trimethylaniline
SMILES CN(C)C1=CC=CC(C)=C1

Specifications

Melting Point -15°C
Density 0.932
Boiling Point 212°C to 215°C
Flash Point 85°C (185°F)
Refractive Index 1.55
Quantity 100 g
UN Number UN1708
Beilstein 1422766
Formula Weight 135.21
Percent Purity 98%
Chemical Name or Material N,N-Dimethyl-m-toluidine
Show More Show Less
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUIDINES, LIQUID
EINECSNumber 204-495-6
RTECSNumber XU5798000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only