Learn More
N,N'-Diethylthiourea, 98%
CAS: 105-55-5 | C5H12N2S | 132.23 g/mol
Supplier: Thermo Scientific Chemicals A171430B
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N,N'-Diethylthiourea | |
| 75°C to 79°C | |
| 186°C (367°F) | |
| MFCD00004925 | |
| 773905 | |
| FLVIGYVXZHLUHP-UHFFFAOYSA-N | |
| 1,3-diethylthiourea | |
| 2735009 | |
| 132.23 |
| 105-55-5 | |
| 1.12 | |
| C5H12N2S | |
| 1000 g | |
| 1,3-diethyl-2-thiourea, n,n'-diethylthiourea, pennzone e, thiourea, n,n'-diethyl, thiate h, n,n'-diethylthiocarbamide, urea, 1,3-diethyl-2-thio, usaf ek-1803, n,n-diethyl-2-thiourea, diethylthiourea | |
| CCNC(=S)NCC | |
| 132.23 | |
| CHEBI:82448 | |
| 98% |
Chemical Identifiers
| 105-55-5 | |
| 132.23 | |
| FLVIGYVXZHLUHP-UHFFFAOYSA-N | |
| 2735009 | |
| 1,3-diethylthiourea |
| C5H12N2S | |
| MFCD00004925 | |
| 1,3-diethyl-2-thiourea, n,n'-diethylthiourea, pennzone e, thiourea, n,n'-diethyl, thiate h, n,n'-diethylthiocarbamide, urea, 1,3-diethyl-2-thio, usaf ek-1803, n,n-diethyl-2-thiourea, diethylthiourea | |
| CHEBI:82448 | |
| CCNC(=S)NCC |
Safety and Handling
GHS H Statement
H318-H351-H302-H317
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause an allergic skin reaction.
P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P314-P330-P333+P313-P363-P501c
H302+H312-H317-H318-H372
EINECSNumber : 203-308-5
RTECSNumber : YS9800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only