missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diethylthiourea, 98%

Catalog No. p-7023515
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1714330 250 g
AAA171430B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1714330 Supplier Thermo Scientific Chemicals Supplier No. A1714330

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 105-55-5 | C5H12N2S | 132.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-55-5
Molecular Formula C5H12N2S
Molecular Weight (g/mol) 132.23
MDL Number MFCD00004925
InChI Key FLVIGYVXZHLUHP-UHFFFAOYSA-N
Synonym 1,3-diethyl-2-thiourea, n,n'-diethylthiourea, pennzone e, thiourea, n,n'-diethyl, thiate h, n,n'-diethylthiocarbamide, urea, 1,3-diethyl-2-thio, usaf ek-1803, n,n-diethyl-2-thiourea, diethylthiourea
PubChem CID 2735009
ChEBI CHEBI:82448
IUPAC Name 1,3-diethylthiourea
SMILES CCNC(=S)NCC

Specifications

Melting Point 75°C to 79°C
Density 1.12
Flash Point 186°C (367°F)
Quantity 250 g
Beilstein 773905
Formula Weight 132.23
Percent Purity 98%
Chemical Name or Material N,N'-Diethylthiourea
Hazard Category H302+H312-H317-H318-H372
Hazard Statement GHS H Statement
H318-H351-H302-H317
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P314-P330-P333+P313-P363-P501c
EINECSNumber 203-308-5
RTECSNumber YS9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only