missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethylglycine Sodium Salt 97.0+%, TCI America™
Supplier: TCI America D050925G
Specifications
| N,N-Diethylglycine Sodium Salt | |
| White-Yellow | |
| MFCD00053497 | |
| XZQDOIKSUNQDGV-UHFFFAOYSA-N | |
| 2-(diethylamino)acetic acid sodium | |
| 23670834 | |
| ≥97.0% (T) |
| 5426-55-1 | |
| C6H13NNaO2 | |
| 25 g | |
| [Na].CCN(CC)CC(O)=O | |
| 154.17 | |
| 153.16 | |
| Crystalline Powder |
Chemical Identifiers
| 5426-55-1 | |
| 154.17 | |
| XZQDOIKSUNQDGV-UHFFFAOYSA-N | |
| 2-(diethylamino)acetic acid sodium |
| C6H13NNaO2 | |
| MFCD00053497 | |
| 23670834 | |
| [Na].CCN(CC)CC(O)=O |
Safety and Handling
TSCA : Yes