missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylglycine Sodium Salt 97.0+%, TCI America™

Catalog No. p-7126258
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D050925G 25 g
D0509100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D050925G Supplier TCI America Supplier No. D050925G
Only null left

Chemical Identifiers

CAS 5426-55-1
Molecular Formula C6H13NNaO2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00053497
InChI Key XZQDOIKSUNQDGV-UHFFFAOYSA-N
PubChem CID 23670834
IUPAC Name 2-(diethylamino)acetic acid sodium
SMILES [Na].CCN(CC)CC(O)=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 153.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diethylglycine Sodium Salt