missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylbenzamide 95.0+%, TCI America™

Supplier:  TCI America D5123100G

 View more versions of this product

Catalog No. D5123100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N,N-Diethylbenzamide
Yellow
C11H15NO
100 g
JLNGEXDJAQASHD-UHFFFAOYSA-N
N,N-diethylbenzamide
15542
≥95.0% (GC)
1696-17-9
280°C
MFCD00026726
rebemid, benzamide, n,n-diethyl, rebemide, benzoyldiethylamine, benzoic acid diethylamide, r 2 insect repellant, benzoic acid n,n-diethylamide, r 2 insect repellent, unii-3y74433zxi, r 2 van
CCN(CC)C(=O)C1=CC=CC=C1
177.25
177.25
Liquid

Chemical Identifiers

1696-17-9
177.25
JLNGEXDJAQASHD-UHFFFAOYSA-N
15542
CCN(CC)C(=O)C1=CC=CC=C1
C11H15NO
MFCD00026726
rebemid, benzamide, n,n-diethyl, rebemide, benzoyldiethylamine, benzoic acid diethylamide, r 2 insect repellant, benzoic acid n,n-diethylamide, r 2 insect repellent, unii-3y74433zxi, r 2 van
N,N-diethylbenzamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : CV4202000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions