missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylbenzamide 95.0+%, TCI America™

Catalog No. p-7128660
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D512325G 25 g
D5123100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D512325G Supplier TCI America Supplier No. D512325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1696-17-9
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD00026726
InChI Key JLNGEXDJAQASHD-UHFFFAOYSA-N
Synonym rebemid, benzamide, n,n-diethyl, rebemide, benzoyldiethylamine, benzoic acid diethylamide, r 2 insect repellant, benzoic acid n,n-diethylamide, r 2 insect repellent, unii-3y74433zxi, r 2 van
PubChem CID 15542
IUPAC Name N,N-diethylbenzamide
SMILES CCN(CC)C(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 280°C
Quantity 25 g
Formula Weight 177.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethylbenzamide
RTECSNumber CV4202000
TSCA Yes